Assay Details
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013091.1 | NP_037223.1 | 6 - 7 | 866 | 71 | |
XM_006237334.2 | XP_006237396.1 | 5 - 6 | 1101 | 71 | ||
GenBank mRNA | AF329976.1 | - | 6 - 7 | 865 | 71 | |
AF329977.1 | - | 6 - 7 | 865 | 71 | ||
AF329978.1 | - | 6 - 7 | 865 | 71 | ||
AF329979.1 | - | 6 - 7 | 865 | 71 | ||
AF329980.1 | - | 6 - 7 | 865 | 71 | ||
AF329981.1 | - | 6 - 7 | 865 | 71 | ||
BC086413.1 | - | 6 - 7 | 886 | 71 | ||
M63122.1 | - | 6 - 7 | 866 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
