Assay Details
Gene Symbol: Il7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013110.2 | NP_037242.2 | 3 - 4 | 242 | 104 | |
XM_006232139.2 | XP_006232201.1 | 3 - 4 | 3088 | 104 | ||
XM_008760860.1 | XP_008759082.1 | 3 - 4 | 3088 | 104 | ||
GenBank mRNA | AF010464.1 | - | 3 - 4 | 251 | 104 | |
AF367210.1 | - | 3 - 4 | 242 | 104 | ||
FQ226549.1 | - | 812 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
