Assay Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013168.2 | NP_037300.2 | 4 - 5 | 332 | 103 | |
XM_006242896.1 | XP_006242958.1 | 4 - 5 | 418 | 103 | ||
XM_006242897.1 | XP_006242959.1 | 4 - 5 | 418 | 103 | ||
GenBank mRNA | BC088162.1 | - | 4 - 5 | 332 | 103 | |
FQ226062.1 | - | 4 - 5 | 255 | 103 | ||
FQ231091.1 | - | 4 - 5 | 316 | 103 | ||
X06827.1 | - | 4 - 5 | 207 | 103 | ||
Y12006.1 | - | 4 - 5 | 202 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
