Assay Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013168.2 | NP_037300.2 | 9 - 10 | 736 | 83 | |
XM_006242896.1 | XP_006242958.1 | 9 - 10 | 822 | 83 | ||
XM_006242897.1 | XP_006242959.1 | 8 - 9 | 744 | 83 | ||
XM_008766148.1 | XP_008764370.1 | 8 - 9 | 906 | 83 | ||
XM_008766149.1 | XP_008764371.1 | 8 - 9 | 843 | 83 | ||
GenBank mRNA | BC088162.1 | - | 9 - 10 | 736 | 7375228 | 83 |
FQ225972.1 | - | 3 - 4 | 221 | 83 | ||
FQ226062.1 | - | 9 - 10 | 659 | 83 | ||
FQ231091.1 | - | 9 - 10 | 83 | |||
FQ234793.1 | - | 1 - 2 | 108 | 83 | ||
FQ234870.1 | - | 7 - 8 | 892 | 83 | ||
X06827.1 | - | 9 - 10 | 611 | 83 | ||
Y12006.1 | - | 9 - 10 | 606 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
