Assay Details
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013174.2 | NP_037306.1 | 1 - 2 | 752 | 124 | |
XM_006240366.1 | XP_006240428.1 | 2 - 3 | 1096 | 124 | ||
GenBank mRNA | BC092195.1 | - | 1 - 2 | 739 | 7313608 | 124 |
U03491.1 | - | 1 - 2 | 752 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
