Assay Details
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013174.2 | NP_037306.1 | 5 - 6 | 1324 | 56 | |
XM_006240366.1 | XP_006240428.1 | 6 - 7 | 1668 | 56 | ||
GenBank mRNA | BC092195.1 | - | 5 - 6 | 1311 | 56 | |
U03491.1 | - | 5 - 6 | 1324 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
