Assay Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146061.1 | NP_001139533.1 | 4 - 5 | 597 | 85 | |
NM_013214.4 | NP_037346.2 | 4 - 5 | 525 | 85 | ||
XM_006239447.2 | XP_006239509.1 | 4 - 5 | 625 | 85 | ||
XM_006239448.1 | XP_006239510.1 | 4 - 5 | 533 | 85 | ||
GenBank mRNA | BC087716.1 | - | 4 - 5 | 477 | 85 | |
D88891.1 | - | 4 - 5 | 597 | 85 | ||
FQ212652.1 | - | 1 - 2 | 76 | 85 | ||
FQ231740.1 | - | 1 - 2 | 76 | 85 | ||
Y09332.1 | - | 4 - 5 | 425 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
