Assay Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146061.1 | NP_001139533.1 | 5 - 6 | 719 | 58 | |
NM_013214.4 | NP_037346.2 | 5 - 6 | 647 | 58 | ||
XM_006239447.2 | XP_006239509.1 | 5 - 6 | 747 | 58 | ||
XM_006239448.1 | XP_006239510.1 | 5 - 6 | 655 | 58 | ||
GenBank mRNA | BC087716.1 | - | 5 - 6 | 599 | 58 | |
D88891.1 | - | 5 - 6 | 719 | 58 | ||
FQ212652.1 | - | 2 - 3 | 198 | 58 | ||
FQ231740.1 | - | 2 - 3 | 198 | 58 | ||
Y09332.1 | - | 5 - 6 | 547 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
