Assay Details
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133316.1 | NP_579850.1 | 8 - 9 | 654 | 64 | |
XM_006238626.2 | XP_006238688.1 | 8 - 9 | 682 | 64 | ||
XM_008763987.1 | XP_008762209.1 | 6 - 7 | 640 | 64 | ||
GenBank mRNA | AF478683.1 | - | 8 - 9 | 654 | 64 | |
BC128728.1 | - | 8 - 9 | 584 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
