Assay Details
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270807.1 | NP_001257736.1 | 4 - 5 | 740 | 66 | |
NM_001270808.1 | NP_001257737.1 | 4 - 5 | 789 | 66 | ||
NM_001270809.1 | NP_001257738.1 | 4 - 5 | 690 | 66 | ||
NM_001270810.1 | NP_001257739.1 | 4 - 5 | 1376 | 66 | ||
NM_001270811.1 | NP_001257740.1 | 4 - 5 | 1726 | 66 | ||
NM_133386.3 | NP_596877.2 | 3 - 4 | 1980 | 66 | ||
XM_006247787.2 | XP_006247849.1 | 4 - 5 | 1060 | 66 | ||
XM_006247788.2 | XP_006247850.1 | 4 - 5 | 571 | 66 | ||
GenBank mRNA | AB049571.1 | - | 4 - 5 | 788 | 66 | |
AB049572.1 | - | 4 - 5 | 740 | 66 | ||
AB049573.1 | - | 4 - 5 | 687 | 66 | ||
AB049574.1 | - | 4 - 5 | 1346 | 66 | ||
AB049575.1 | - | 3 - 4 | 1493 | 66 | ||
BC081738.1 | - | 3 - 4 | 1945 | 66 | ||
DQ486894.1 | - | 4 - 5 | 528 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
