Assay Details
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133420.1 | NP_596911.1 | 6 - 7 | 1710 | 62 | |
XM_008770735.1 | XP_008768957.1 | 6 - 7 | 1552 | 62 | ||
GenBank mRNA | AY574253.1 | - | 5 - 6 | 1406 | 62 | |
L10077.1 | - | 6 - 7 | 1710 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
