Assay Details
Gene Symbol: Mob4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133528.1 | NP_598212.1 | 2 - 3 | 129 | 103 | |
GenBank mRNA | AJ132008.1 | - | 2 - 3 | 129 | 103 | |
BC085708.1 | - | 2 - 3 | 150 | 103 | ||
FQ229796.1 | - | 2 - 3 | 145 | 102 | ||
FQ233766.1 | - | 2 - 3 | 152 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
