Assay Details
Gene Symbol: Mob4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133528.1 | NP_598212.1 | 4 - 5 | 273 | 87 | |
GenBank mRNA | AJ132008.1 | - | 4 - 5 | 273 | 87 | |
BC085708.1 | - | 4 - 5 | 294 | 87 | ||
FQ233766.1 | - | 4 - 5 | 296 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
