Assay Details
Gene Symbol: Stau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007149.1 | NP_001007150.1 | 7 - 8 | 1334 | 81 | |
NM_001007150.1 | NP_001007151.1 | 5 - 6 | 1061 | 81 | ||
NM_134466.2 | NP_604461.2 | 6 - 7 | 1157 | 81 | ||
GenBank mRNA | AF483620.1 | - | 7 - 8 | 1334 | 81 | |
AF483621.1 | - | 7 - 8 | 1334 | 81 | ||
AY549445.1 | - | 5 - 6 | 1061 | 81 | ||
AY549446.1 | - | 5 - 6 | 1061 | 81 | ||
AY549447.1 | - | 6 - 7 | 1157 | 81 | ||
AY549448.1 | - | 6 - 7 | 1157 | 81 | ||
AY684789.1 | - | 1061 | 81 | |||
AY684790.1 | - | 1061 | 81 | |||
BC085705.1 | - | 1693 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
