Assay Details
Gene Symbol: Mgll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138502.2 | NP_612511.1 | 4 - 5 | 485 | 63 | |
XM_008763053.1 | XP_008761275.1 | 4 - 5 | 1143 | 63 | ||
GenBank mRNA | AY081195.1 | - | 3 - 4 | 368 | 63 | |
BC107920.1 | - | 4 - 5 | 485 | 63 | ||
FQ227013.1 | - | 4 - 5 | 1111 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
