Assay Details
Gene Symbol: Ttl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138536.1 | NP_612545.1 | 3 - 4 | 613 | 76 | |
XM_006234946.2 | XP_006235008.1 | 3 - 4 | 686 | 76 | ||
GenBank mRNA | AF207605.1 | - | 3 - 4 | 613 | 76 | |
U53214.1 | - | 3 - 4 | 466 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
