Assay Details
Gene Symbol: Epcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138541.1 | NP_612550.1 | 4 - 5 | 561 | 76 | |
GenBank mRNA | AJ001044.1 | - | 4 - 5 | 616 | 76 | |
BC061787.1 | - | 4 - 5 | 565 | 76 | ||
BC072691.1 | - | 4 - 5 | 585 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
