Assay Details
Gene Symbol: Slc2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138827.1 | NP_620182.1 | 4 - 5 | 726 | 64 | |
GenBank mRNA | BC061873.1 | - | 4 - 5 | 654 | 64 | |
M13979.1 | - | 4 - 5 | 726 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
