Assay Details
Gene Symbol: Snrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138833.1 | NP_620188.1 | 4 - 5 | 1089 | 58 | |
XM_006244057.2 | XP_006244119.1 | 5 - 6 | 1135 | 58 | ||
XM_006244058.2 | XP_006244120.1 | 5 - 6 | 1169 | 58 | ||
XM_006244059.1 | XP_006244121.1 | 5 - 6 | 1128 | 58 | ||
XM_006244060.2 | XP_006244122.1 | 4 - 5 | 1143 | 58 | ||
XM_006244061.1 | XP_006244123.1 | 4 - 5 | 1149 | 58 | ||
XM_006244062.2 | XP_006244124.1 | 4 - 5 | 1077 | 58 | ||
XM_008766666.1 | XP_008764888.1 | 4 - 5 | 1068 | 58 | ||
XM_008766667.1 | XP_008764889.1 | 4 - 5 | 1210 | 58 | ||
GenBank mRNA | X89383.1 | - | 4 - 5 | 1089 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
