Assay Details
Gene Symbol: Gpm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138846.1 | NP_620201.1 | 3 - 4 | 643 | 61 | |
XM_006256849.2 | XP_006256911.1 | 4 - 5 | 1182 | 61 | ||
XM_006256850.2 | XP_006256912.1 | 4 - 5 | 1184 | 61 | ||
XM_006256851.2 | XP_006256913.1 | 3 - 4 | 706 | 61 | ||
XM_008773151.1 | XP_008771373.1 | 3 - 4 | 695 | 61 | ||
GenBank mRNA | AB036421.1 | - | 3 - 4 | 643 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
