Assay Details
Gene Symbol: Ptgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138863.2 | NP_620218.1 | 2 - 3 | 148 | 65 | |
GenBank mRNA | BC089775.1 | - | 2 - 3 | 148 | 65 | |
FQ218841.1 | - | 2 - 3 | 167 | 65 | ||
FQ221589.1 | - | 2 - 3 | 164 | 65 | ||
FQ229147.1 | - | 2 - 3 | 164 | 65 | ||
FQ229679.1 | - | 2 - 3 | 164 | 65 | ||
U66322.2 | - | 1 - 2 | 113 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
