Assay Details
Gene Symbol: Pla1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138882.1 | NP_620237.1 | 5 - 6 | 673 | 68 | |
XM_006248377.2 | XP_006248439.1 | 5 - 6 | 1082 | 68 | ||
GenBank mRNA | BC078727.1 | - | 5 - 6 | 673 | 68 | |
D88666.1 | - | 5 - 6 | 673 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
