Assay Details
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138884.1 | NP_620239.1 | 6 - 7 | 729 | 94 | |
XM_006236285.1 | XP_006236347.1 | 6 - 7 | 746 | 94 | ||
GenBank mRNA | D17309.1 | - | 6 - 7 | 729 | 94 | |
FQ211413.1 | - | 6 - 7 | 898 | 94 | ||
FQ219185.1 | - | 6 - 7 | 745 | 94 | ||
FQ219730.1 | - | 6 - 7 | 689 | 95 | ||
S80431.1 | - | 6 - 7 | 701 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
