Assay Details
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139080.2 | NP_620780.2 | 7 - 8 | 1206 | 67 | |
GenBank mRNA | AB066102.1 | - | 6 - 7 | 1190 | 67 | |
BC078718.1 | - | 7 - 8 | 1206 | 7115290 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
