Assay Details
Gene Symbol: Xrcc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139080.2 | NP_620780.2 | 8 - 9 | 1370 | 75 | |
GenBank mRNA | AB066102.1 | - | 7 - 8 | 1354 | 75 | |
BC078718.1 | - | 8 - 9 | 1370 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
