Assay Details
Gene Symbol: Slc25a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270788.1 | NP_001257717.1 | 4 - 5 | 546 | 107 | |
NM_139100.2 | NP_620800.2 | 4 - 5 | 543 | 107 | ||
GenBank mRNA | BC070918.1 | - | 4 - 5 | 485 | 107 | |
CB577607.1 | - | 4 - 5 | 543 | 107 | ||
FQ210190.1 | - | 4 - 5 | 497 | 107 | ||
FQ213750.1 | - | 4 - 5 | 502 | 107 | ||
FQ214698.1 | - | 4 - 5 | 500 | 107 | ||
FQ215315.1 | - | 4 - 5 | 500 | 107 | ||
FQ215697.1 | - | 4 - 5 | 500 | 107 | ||
FQ215957.1 | - | 4 - 5 | 500 | 107 | ||
FQ216361.1 | - | 4 - 5 | 500 | 107 | ||
FQ216409.1 | - | 4 - 5 | 507 | 107 | ||
FQ216421.1 | - | 4 - 5 | 500 | 107 | ||
FQ216474.1 | - | 4 - 5 | 507 | 107 | ||
FQ217707.1 | - | 4 - 5 | 500 | 107 | ||
FQ220537.1 | - | 4 - 5 | 497 | 107 | ||
FQ223916.1 | - | 4 - 5 | 500 | 107 | ||
FQ227446.1 | - | 4 - 5 | 497 | 107 | ||
FQ229113.1 | - | 4 - 5 | 497 | 107 | ||
FQ232293.1 | - | 2 - 3 | 302 | 107 | ||
FQ232835.1 | - | 4 - 5 | 493 | 107 | ||
FQ234854.1 | - | 4 - 5 | 497 | 107 | ||
M23984.1 | - | 3 - 4 | 462 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
