Assay Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248989.2 | XP_006249051.1 | 9 - 10 | 3418 | 83 | |
GenBank mRNA | AF030243.1 | - | 4 - 5 | 622 | 83 | |
BP475867.1 | - | 3 - 4 | 83 | |||
FQ214452.1 | - | 6 - 7 | 996 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
