Assay Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139336.1 | NP_647552.1 | 2 - 3 | 169 | 152 | |
XM_008766750.1 | XP_008764972.1 | 2 - 3 | 214 | 152 | ||
GenBank mRNA | AF482705.1 | - | 2 - 3 | 169 | 152 | |
BC086988.1 | - | 2 - 3 | 180 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
