Assay Details
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139342.1 | NP_647558.1 | 3 - 4 | 474 | 64 | |
XM_006251928.2 | XP_006251990.1 | 2 - 3 | 1326 | 64 | ||
XM_008770661.1 | XP_008768883.1 | 1 - 2 | 287 | 64 | ||
XM_008770662.1 | XP_008768884.1 | 2 - 3 | 658 | 64 | ||
GenBank mRNA | AF036537.2 | - | 3 - 4 | 474 | 64 | |
BC158580.1 | - | 1 - 2 | 41 | 7378607 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF036537.2, BC158580.1, NM_139342.1, XM_006251928.2, XM_008770661.1, XM_008770662.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
