Assay Details
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145092.2 | NP_659560.2 | 6 - 7 | 980 | 69 | |
XM_006250027.2 | XP_006250089.1 | 6 - 7 | 525 | 69 | ||
XM_006250028.2 | XP_006250090.1 | 6 - 7 | 1111 | 69 | ||
GenBank mRNA | BC086987.1 | - | 6 - 7 | 980 | 69 | |
U19614.1 | - | 5 - 6 | 769 | 69 | ||
U20286.1 | - | 6 - 7 | 437 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
