Assay Details
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145092.2 | NP_659560.2 | 7 - 8 | 1031 | 82 | |
XM_006250027.2 | XP_006250089.1 | 7 - 8 | 576 | 82 | ||
XM_006250028.2 | XP_006250090.1 | 7 - 8 | 1162 | 82 | ||
GenBank mRNA | BC086987.1 | - | 7 - 8 | 1031 | 7129718 | 82 |
U19614.1 | - | 6 - 7 | 820 | 82 | ||
U20286.1 | - | 7 - 8 | 488 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
