Assay Details
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164073.1 | NP_001157545.1 | 5 - 6 | 527 | 133 | |
NM_145788.2 | NP_665731.2 | 5 - 6 | 538 | 133 | ||
XM_006234273.2 | XP_006234335.1 | 5 - 6 | 600 | 133 | ||
XM_006234274.2 | XP_006234336.1 | 5 - 6 | 586 | 133 | ||
GenBank mRNA | AJ422211.1 | - | 4 - 5 | 398 | 133 | |
BC088854.1 | - | 6 - 7 | 606 | 133 | ||
BC129086.1 | - | 5 - 6 | 527 | 133 | ||
EV776506.1 | - | 5 - 6 | 538 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
