Assay Details
Gene Symbol: Ppm1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147209.2 | NP_671742.1 | 4 - 5 | 535 | 88 | |
GenBank mRNA | AF525687.1 | - | 4 - 5 | 411 | 88 | |
BC062083.1 | - | 4 - 5 | 535 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
