Assay Details
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153311.1 | NP_695223.1 | 1 - 2 | 77 | 81 | |
XM_008766194.1 | XP_008764416.1 | 1 - 2 | 162 | 81 | ||
GenBank mRNA | AF537098.1 | - | 1 - 2 | 77 | 81 | |
AF537099.1 | - | 1 - 2 | 77 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF537098.1, AF537099.1, NM_153311.1, XM_008766194.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
