Assay Details
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172224.1 | NP_757378.1 | 1 - 2 | 281 | 84 | |
GenBank mRNA | AY160191.1 | - | 1 - 2 | 281 | 84 | |
BC083544.1 | - | 1 - 2 | 255 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
