Assay Details
Gene Symbol: Elavl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172324.1 | NP_758827.1 | 4 - 5 | 528 | 82 | |
XM_006242616.2 | XP_006242678.1 | 4 - 5 | 872 | 82 | ||
XM_008765920.1 | XP_008764142.1 | 4 - 5 | 715 | 82 | ||
GenBank mRNA | AB097198.1 | - | 4 - 5 | 528 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
