Assay Details
Gene Symbol: Zp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172330.3 | NP_758833.2 | 4 - 5 | 607 | 57 | |
GenBank mRNA | AF456325.1 | - | 4 - 5 | 608 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
