Assay Details
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172334.2 | NP_758837.1 | 1 - 2 | 106 | 70 | |
XM_006230701.2 | XP_006230763.1 | 1 - 2 | 162 | 70 | ||
GenBank mRNA | AY093421.1 | - | 1 - 2 | 107 | 70 | |
BC061802.1 | - | 1 - 2 | 127 | 70 | ||
FQ228181.1 | - | 1 - 2 | 158 | 70 | ||
FQ234469.1 | - | 1 - 2 | 158 | 70 | ||
FQ234564.1 | - | 1 - 2 | 158 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
