Assay Details
Gene Symbol: Dpy30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170545.1 | NP_001164016.1 | 2 - 3 | 141 | 78 | |
NM_173117.2 | NP_775140.2 | 2 - 3 | 180 | 78 | ||
GenBank mRNA | AY129401.1 | - | 2 - 3 | 180 | 78 | |
FM121127.1 | - | 2 - 3 | 147 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY129401.1, NM_001170545.1, NM_173117.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
