Assay Details
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012565.2 | NP_036697.1 | 1 - 2 | 153 | 62 | |
GenBank mRNA | J04218.1 | - | 1 - 2 | 112 | 62 | |
X53588.1 | - | 1 - 2 | 136 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
