Assay Details
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270850.1 | NP_001257779.1 | 2 - 3 | 246 | 64 | |
NM_012565.2 | NP_036697.1 | 2 - 3 | 318 | 64 | ||
XM_006251179.2 | XP_006251241.1 | 2 - 3 | 1163 | 64 | ||
GenBank mRNA | J04218.1 | - | 2 - 3 | 277 | 64 | |
KJ026953.1 | - | 2 - 3 | 210 | 64 | ||
M25807.1 | - | 2 - 3 | 246 | 64 | ||
M58759.1 | - | 2 - 3 | 700 | 64 | ||
X53588.1 | - | 2 - 3 | 301 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
