Assay Details
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270849.1 | NP_001257778.1 | 4 - 5 | 569 | 70 | |
NM_012565.2 | NP_036697.1 | 3 - 4 | 470 | 70 | ||
XM_006251179.2 | XP_006251241.1 | 3 - 4 | 1315 | 70 | ||
GenBank mRNA | J04218.1 | - | 3 - 4 | 429 | 70 | |
KJ026953.1 | - | 3 - 4 | 362 | 70 | ||
M58759.1 | - | 3 - 4 | 852 | 70 | ||
X53588.1 | - | 3 - 4 | 453 | 70 | ||
X53589.1 | - | 4 - 5 | 569 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
