Assay Details
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270849.1 | NP_001257778.1 | 9 - 10 | 1227 | 61 | |
NM_001270850.1 | NP_001257779.1 | 8 - 9 | 1005 | 61 | ||
NM_012565.2 | NP_036697.1 | 8 - 9 | 1128 | 61 | ||
XM_006251179.2 | XP_006251241.1 | 8 - 9 | 1973 | 61 | ||
GenBank mRNA | J04218.1 | - | 8 - 9 | 1087 | 61 | |
KJ026953.1 | - | 8 - 9 | 1020 | 61 | ||
M25807.1 | - | 8 - 9 | 1005 | 61 | ||
X53588.1 | - | 8 - 9 | 1111 | 61 | ||
X53589.1 | - | 9 - 10 | 1227 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
