Assay Details
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012571.2 | NP_036703.2 | 3 - 4 | 508 | 59 | |
GenBank mRNA | BC061877.1 | - | 3 - 4 | 481 | 59 | |
CB578328.1 | - | 1 - 2 | 72 | 59 | ||
D00252.1 | - | 3 - 4 | 465 | 59 | ||
FM055803.1 | - | 3 - 4 | 430 | 59 | ||
J04171.1 | - | 3 - 4 | 449 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
