Assay Details
Gene Symbol: Got1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012571.2 | NP_036703.2 | 7 - 8 | 1042 | 61 | |
GenBank mRNA | BC061877.1 | - | 7 - 8 | 1015 | 61 | |
D00252.1 | - | 7 - 8 | 999 | 61 | ||
FN802619.1 | - | 1 - 2 | 87 | 61 | ||
FQ219727.1 | - | 4 - 5 | 530 | 61 | ||
J04171.1 | - | 7 - 8 | 983 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
