Assay Details
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012598.2 | NP_036730.1 | 7 - 8 | 1332 | 86 | |
GenBank mRNA | BC081836.1 | - | 7 - 8 | 1332 | 86 | |
L03294.1 | - | 1317 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
