Assay Details
Gene Symbol: Mylpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012605.2 | NP_036737.1 | 6 - 7 | 445 | 55 | |
GenBank mRNA | BI278596.1 | - | 1 - 2 | 55 | ||
FQ216934.1 | - | 6 - 7 | 461 | 55 | ||
FQ217090.1 | - | 6 - 7 | 463 | 55 | ||
FQ217098.1 | - | 6 - 7 | 461 | 55 | ||
FQ217115.1 | - | 4 - 5 | 310 | 55 | ||
FQ217155.1 | - | 6 - 7 | 461 | 55 | ||
FQ217219.1 | - | 6 - 7 | 461 | 55 | ||
FQ217259.1 | - | 6 - 7 | 461 | 55 | ||
FQ217349.1 | - | 5 - 6 | 385 | 55 | ||
FQ217365.1 | - | 6 - 7 | 460 | 55 | ||
FQ217368.1 | - | 6 - 7 | 477 | 55 | ||
FQ217533.1 | - | 6 - 7 | 489 | 55 | ||
FQ217703.1 | - | 6 - 7 | 461 | 55 | ||
FQ217740.1 | - | 6 - 7 | 477 | 55 | ||
FQ217876.1 | - | 1 - 2 | 43 | 55 | ||
FQ217881.1 | - | 6 - 7 | 458 | 55 | ||
FQ217944.1 | - | 6 - 7 | 461 | 55 | ||
FQ218017.1 | - | 6 - 7 | 461 | 55 | ||
FQ223762.1 | - | 6 - 7 | 442 | 55 | ||
FQ223868.1 | - | 6 - 7 | 461 | 55 | ||
FQ223911.1 | - | 6 - 7 | 477 | 55 | ||
FQ223944.1 | - | 5 - 6 | 327 | 55 | ||
FQ223946.1 | - | 6 - 7 | 461 | 55 | ||
FQ223966.1 | - | 6 - 7 | 461 | 55 | ||
FQ223985.1 | - | 6 - 7 | 461 | 55 | ||
FQ224150.1 | - | 4 - 5 | 304 | 55 | ||
FQ224231.1 | - | 6 - 7 | 461 | 55 | ||
FQ224253.1 | - | 6 - 7 | 460 | 55 | ||
FQ224391.1 | - | 6 - 7 | 459 | 55 | ||
FQ224404.1 | - | 6 - 7 | 459 | 55 | ||
FQ224511.1 | - | 6 - 7 | 461 | 55 | ||
FQ224525.1 | - | 6 - 7 | 480 | 55 | ||
FQ224781.1 | - | 6 - 7 | 477 | 55 | ||
J00754.1 | - | 3 - 4 | 197 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
