Assay Details
Gene Symbol: Hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012669.1 | NP_036801.1 | 3 - 4 | 862 | 65 | |
GenBank mRNA | J03170.1 | - | 3 - 4 | 862 | 65 | |
X53297.1 | - | 2 - 3 | 220 | 65 | ||
X54423.1 | - | 3 - 4 | 712 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
