Assay Details
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012754.1 | NP_036886.1 | 6 - 7 | 1365 | 92 | |
GenBank mRNA | AB190769.1 | - | 5 - 6 | 1188 | 92 | |
AF042058.1 | - | 5 - 6 | 949 | 92 | ||
AF042060.1 | - | 4 - 5 | 832 | 92 | ||
AF161187.1 | - | 3 - 4 | 361 | 92 | ||
U57439.1 | - | 6 - 7 | 1365 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
