Assay Details
Gene Symbol: Acadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012819.1 | NP_036951.1 | 10 - 11 | 1210 | 60 | |
GenBank mRNA | BC062006.1 | - | 10 - 11 | 1208 | 60 | |
J05029.1 | - | 10 - 11 | 1221 | 60 | ||
L11276.1 | - | 10 - 11 | 1210 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
carnitine catabolic process
long-chain fatty acid catabolic process
cellular lipid catabolic process
negative regulation of fatty acid biosynthetic process
negative regulation of fatty acid oxidation
protein homotetramerization
regulation of cholesterol metabolic process
fatty-acyl-CoA binding
long-chain-acyl-CoA dehydrogenase activity
palmitoyl-CoA oxidase activity
flavin adenine dinucleotide binding
temperature homeostasis
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
carnitine catabolic process
long-chain fatty acid catabolic process
cellular lipid catabolic process
negative regulation of fatty acid biosynthetic process
negative regulation of fatty acid oxidation
protein homotetramerization
regulation of cholesterol metabolic process
fatty-acyl-CoA binding
long-chain-acyl-CoA dehydrogenase activity
palmitoyl-CoA oxidase activity
flavin adenine dinucleotide binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110543 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14184 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14184 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14184 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14184 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700182615 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66870 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103581 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27469 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23707 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700182615 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10928613 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928610 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928611 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928603 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928608 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928604 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928612 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928606 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928605 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928607 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928609 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10928602 | - | - |
Rat Genome 230 2.0 Array | 1367735_at | - | - |
Rat Genome 230 2.0 Array | 1367735_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367735_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367735_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0900017574.rn.1 | TC0900002139.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900017575.rn.1 | TC0900002139.rn.1 | RTA TC |
Rat Expression Set 230 | 1367735_at | - | - |
Rat Expression Set 230 | 1367735_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10928602 | - | - |
Rat Genome U34 Set | J05029_s_at | - | - |
Rat Genome U34 Set | rc_AA848753_at | - | - |
Rat Genome U34 Set | J05029_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21359296 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355733 | - | - |
RatRef-12 Expression BeadChip | ILMN_69215 | - | - |
